missing translation for 'onlineSavingsMsg'
Learn More

Chem-Lab 500ML Lithium standard solution (Spectro ECON)(5.33 g Li2CO3 / l 2 % HNO3), for AAS

Brand:  Chem-Lab CL01.1217.0500


500ML Lithium standard solution (Spectro ECON)(5.33 g Li2CO3 / l 2 % HNO3), for AAS

Product Code. 11999082

  • ONLINE EXCLUSIVE PRICE
    49.00€
    23.23€ / 500mL
    Save 25.77€
    52% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lithium standard solution 1000 µg/ml

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Nitric acid, Lithiumcarbonate

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.