Learn More
Spectrum™ Labs Spectra/Por™ 6 15 kD MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
Brand:  Spectrum™ 132564T
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15350722
  • 127.00€ / 1m
Estimated Shipment: 23-08-2024
to see stock.
Includes:
  • 1 meter of Spectra/Por 6 Dialysis Tubing
  • 1 Standard Spectra/Por Closure (for buoyancy)
  • 1 Weighted Spectra/Por Closure (for orientation)
  • 5 Tubing Opening Picks
Description
Description
  • Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
  • Membrane packaged wet in preservative solution of 0.05% sodium azide
  • Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
  • Include a pair of closures and convenient tubing opening picks
Specifications
Specifications
15 kDa
20.4 mm
3.3 mL/cm vol./length
1 m Tubing/Kit