missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Phosphor Bronze 5% A, CDA 510, C51000, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Phosphor Bronze 5% A, CDA 510, C51000, 50g
Specifications
Specifications
| Chemical Name or Material | Arsenic/Antimony/Iron/Lead/Silicon/Zinc/Nickel/Phosphorus/Tin/Copper/Aluminium/Manganese/Carbon |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.0011 %/Antimony 0.0012 %/Iron 0.0013 %/Lead 0.0045 %/Silicon 0.0046 %/Zinc 0.007 %/Nickel 0.009 %/Phosphorus 0.196 %/Tin 4.55 %/Copper 95.22 %/Aluminium (<0.001) %/Manganese (0.0007) %/Carbon (0.0014) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction