Learn More
1,2-Bis(2,4,6-tribromophenoxy)ethane-d4, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 1,2-Bis(2,4,6-tribromophenoxy)ethane-d4 |
| CAS | 1794620-64-6 |
| Molecular Formula | C14H4D4Br6O2 |
| Quantity | 50 mg |
| Synonym | 1,1'-[1,2-(Ethane-d4)diylbis(oxy)]bis[2,4,6-tribromobenzene, BTBPE-d4, FF 680-d4, FI 680-d4, FM 680-d4, FireMaster 680-d4, FireMaster FF 680-d4 |
| InChI Formula | InChI=1S/C14H8Br6O2/c15-7-3-9(17)13(10(18)4-7)21-1-2-22-14-11(19)5-8(16)6-12(14)20/h3-6H,1-2H2/i1D2,2D2 |
| SMILES | [2H]C([2H])(Oc1c(Br)cc(Br)cc1Br)C([2H])([2H])Oc2c(Br)cc(Br)cc2Br |
| IUPAC Name | 1,3,5-tribromo-2-[1,1,2,2-tetradeuterio-2-(2,4,6-tribromophenoxy)ethoxy]benzene |
| Molecular Weight (g/mol) | 691.66 |
| Formula Weight | 685.5876 |