missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-2-[[4-[[(2-Amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]methylamino]benzoyl]amino]pentanedioic Acid (N-Methylfolic Acid, Methopterin), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
(2S)-2-[[4-[[(2-Amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]methylamino]benzoyl]amino]pentanedioic Acid (N-Methylfolic Acid, Methopterin)
Specifications
Specifications
| Chemical Name or Material | Methopteridine |
| CAS | 2410-93-7 |
| Analyte or Component Names | Methopteridine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H21 N7 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methotrexate Imp. C (EP), Methotrexate USP Related Compound C, Methotrexate USP RC C, N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic Acid, Glutamic acid, N-[p-[[(2-amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]methylamino]benzoyl]-, L- (7CI,8CI), Glutamic acid, N-[p-[[(2-amino-4-hydroxy-6-pteridinyl)methyl]methylamino]benzoyl]- (6CI), L-Glutamic acid, N-[4-[[(2-amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl]methylamino]benzoyl]- (9CI), 10-Methylfolic acid, 10-Methylpteroylglutamic acid, Methopterin, Methopterine, N10-Methylfolic acid, N10-Methylpteroylglutamic acid, NSC 107144, Methotrexate Related Compound C, Methotrexate Impurity C |
| InChI Formula | InChI=1S/C20H21N7O6/c1-27(9-11-8-22-16-15(23-11)18(31)26-20(21)25-16)12-4-2-10(3-5-12)17(30)24-13(19(32)33)6-7-14(28)29/h2-5,8,13H,6-7,9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H3,21,22,25,26,31)/t13-/m0/s1 |
| SMILES | CN(Cc1cnc2NC(=NC(=O)c2n1)N)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O |
| IUPAC Name | (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu