Learn More
3-Methylbutyraldehyde-2,2-d2, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 3-Methylbutyraldehyde-2,2-d2 |
| CAS | 352431-47-1 |
| Molecular Formula | C5 D2 H8 O |
| Quantity | 50 mg |
| Synonym | Butanal-2,2-d2','3-methyl- (9CI), 3-Methylbutyraldehyde-2,2-d2, 3-Methylbutyraldehyde-2,2-D2, 3-Methylbutanal-2,2-d2 |
| InChI Formula | InChI=1S/C5H10O/c1-5(2)3-4-6/h4-5H,3H2,1-2H3/i3D2 |
| SMILES | [2H]C([2H])(C=O)C(C)C |
| IUPAC Name | 2,2-dideuterio-3-methylbutanal |
| Molecular Weight (g/mol) | 88.145 |
| Formula Weight | 88.086 |