missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-N-propyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxamide Hydrochloride (Articaine Acid Propionamide Hydrochloride), Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
4-Methyl-N-propyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxamide Hydrochloride (Articaine Acid Propionamide Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Articaine Acid Propionamide Hydrochloride |
| CAS | 2983985-81-3 |
| Analyte or Component Names | Articaine Acid Propionamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H25 N3 O2 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Articaine Hydrochloride Imp. F (EP), Articaine Imp. F (EP), 4-Methyl-N-propyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxamide Hydrochloride, Articaine Hydrochloride Imp. F (EP) as Hydrochloride, Articaine Acid Propionamide Hydrochloride, Articaine Hydrochloride Impurity F as Hydrochloride, Articaine Impurity F as Hydrochloride |
| InChI Formula | InChI=1S/C4H3N5O.H2O/c10-4-2-3(6-1-5-2)7-9-8-4;/h1H,(H2,5,6,7,8,10);1H5 |
| SMILES | Cl.CCCNC(C)C(=O)Nc1c(C)csc1C(=O)NCCC |
| IUPAC Name | 4-methyl-N-propyl-3-[2-(propylamino)propanoylamino]thiophene-2-carboxamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction