Learn More
Amlodipine-d4, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Amlodipine-d4 |
| CAS | 1185246-14-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20H21D4ClN2O5 |
| Quantity | 50 mg |
| Synonym | 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl)-6-methyl-1,4-dihydropyridine-d4, (R,S)-Amlodipine-d4, Amlopres-d4, Intervask-d4, Pelmec-d4, UK-48340-d4 |
| InChI Formula | InChI=1S/C20H25ClN2O5/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21/h5-8,17,23H,4,9-11,22H2,1-3H3/i9D2,10D2 |
| SMILES | NC([2H])([2H])C([2H])([2H])OCC1=C(C(OCC)=O)C(C2=CC=CC=C2Cl)C(C(OC)=O)=C(C)N1 |
| IUPAC Name | 3-O-ethyl 5-O-methyl 2-[(2-amino-1,1,2,2-tetradeuterioethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Molecular Weight (g/mol) | 412.9 |
| Show More |