Learn More
L-Valine-d8, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Valine d8 |
| CAS | 35045-72-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5 2H8 H3 N O2 |
| Quantity | 50 mg |
| Synonym | L-Valine-2,3,4,4,4,4',4',4'-d8, Butanoic-2,3,4,4,4-d5 acid','2-amino-3-(methyl-d3)-','(S)-, Valine-2,3,4,4,4,4',4',4'-d8','L- (8CI), L-Valine-d8, L-Valine-2,3,4,4,4,4',4',4'-D8, (2S)-2-Amino-3-(methyl-d3)butanoic-2,3,4,4,4-d5 acid |
| InChI Formula | InChI=1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i1D3,2D3,3D,4D |
| SMILES | [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])[C@]([2H])(N)C(=O)O |
| IUPAC Name | (2S)-2-amino-2,3,4,4,4-pentadeuterio-3-(trideuteriomethyl)butanoic acid |
| Molecular Weight (g/mol) | 125.2 |
| Show More |