missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-(2-carboxypropyl)-L-cysteine Bis(dicyclohexylammonium) Salt(Mixture of Diastereomers), TRC

Product Code. 30325388
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30325388

Brand: Toronto Research Chemicals TRCA17201050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-S-(2-carboxypropyl)-L-cysteine Bis(dicyclohexylammonium) Salt(Mixture of Diastereomers)
Molecular Formula C9H15NO5S . 2(C12H23N)
Quantity 50 mg
Synonym N-Acetyl-3-(2-carboxypropyl)thio]alanine Bis(dicyclohexylammonium) Salt
InChI Formula InChI=1S/2C12H23N.C9H15NO5S/c2*1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(8(12)13)3-16-4-7(9(14)15)10-6(2)11/h2*11-13H,1-10H2;5,7H,3-4H2,1-2H3,(H,10,11)(H,12,13)(H,14,15)/t;;5?,7-/m..0/s1
SMILES CC(CSC[C@H](NC(=O)C)C(=O)O)C(=O)O.C1CCC(CC1)NC2CCCCC2.C3CCC(CC3)NC4CCCCC4
IUPAC Name 3-[(2R)-2-acetamido-2-carboxy-ethyl]sulfanyl-2-methyl-propanoic acid;N-cyclohexylcyclohexanamine
Molecular Weight (g/mol) 611.91954
Formula Weight 611.4331
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.