missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Hydroxy Vildagliptin, TRC
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Specifications
Specifications
| Chemical Name or Material | N-Hydroxy Vildagliptin |
| Analyte or Component Names | N-Hydroxy Vildagliptin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H25 N3 O3 |
| InChI Formula | InChI=1S/C17H25N3O3/c18-9-14-2-1-3-19(14)15(21)10-20(23)16-5-12-4-13(6-16)8-17(22,7-12)11-16/h12-14,22-23H,1-8,10-11H2/t12-,13+,14-,16+,17-/m0/s1 |
| Solution Type | Neat |
| SMILES | ON(CC(=O)N1CCC[C@H]1C#N)[C@@]23C[C@@H]4C[C@@H](C[C@@](O)(C4)C2)C3 |
| IUPAC Name | (2S)-1-[2-[hydroxy-[(5S,7R)-3-hydroxy-1-adamantyl]amino]acetyl]pyrrolidine-2-carbonitrile |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 319.399 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction