missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis[2-[[[2-[(dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
N,N'-Bis[2-[[[2-[(dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis[2-[[[2-[(dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine |
| CAS | 1193434-63-7 |
| Analyte or Component Names | N,N'-Bis[2-[[[2-[(dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H33 N7 O2 S4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N,N'-Bis[2-[[[2-[(dimethylamino)methyl]thiazol-4-yl]methyl]sulphanyl]ethyl]-2-nitroethene-1,1-diamine, Nizatidine Imp. G (EP), Nizatidine Impurity G |
| InChI Formula | InChI=1S/C20H33N7O2S4/c1-25(2)10-19-23-16(14-32-19)12-30-7-5-21-18(9-27(28)29)22-6-8-31-13-17-15-33-20(24-17)11-26(3)4/h9,14-15,21-22H,5-8,10-13H2,1-4H3 |
| SMILES | CN(C)Cc1nc(CSCCNC(=C[N+](=O)[O-])NCCSCc2csc(CN(C)C)n2)cs1 |
| IUPAC Name | 1-N,1-N'-bis[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfanyl]ethyl]-2-nitroethene-1,1-diamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction