missing translation for 'onlineSavingsMsg'
Learn More

Testosterone 17-phenylpropionate, Lipomed

Product Code. 30460885
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30460885

Brand: Lipomed LPMTES124150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Testosterone 17-phenylpropionate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Phenylpropionate
CAS 1255-49-8
Analyte or Component Names Testosterone Phenylpropionate
Shipping Condition Room Temperature
Molecular Formula C28 H36 O3
Synonym Testosterone, hydrocinnamate (6 CI,7 CI,8 CI), Hydrocinnamic acid, ester with testosterone (8 CI), Androject, Hydrocinnamic acid, testosterone ester, NSC 26643, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate, 3-Oxoandrost-4-en-17 beta-yl beta-phenylpropionate
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES C[C@]12 CC[C@H]3 [C@@H](CCC4=CC(=O)CC[C@]34 C)[C@@H]1 CC[C@@H]2 OC(=O)CCc5ccccc5
IUPAC Name [(8 R,9 S,10 R,13 S,14 S,17 S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-phenylpropanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 420.58
Quantity 50 mg
Formula Weight 420.2664 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.