missing translation for 'onlineSavingsMsg'
Learn More

Zopiclone, Lipomed

Product Code. 30462277
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 30462277

Brand: Lipomed LPMZOP921FB50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Zopiclone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zopiclone
CAS 43200-80-2
Analyte or Component Names Zopiclone
Shipping Condition Room Temperature
Molecular Formula C17 H17 Cl N6 O3
Synonym Zopiclone
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN1CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4
IUPAC Name [6-(5-chloropyridin-2-yl)-5-oxo-7 H-pyrrolo[3,4-b]pyrazin-7-yl] 4-methylpiperazine-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 388.81
Quantity 50 mg
Formula Weight 388.1051 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.