missing translation for 'onlineSavingsMsg'
Learn More

Bromoethane-d5, TRC

Product Code. 30342287
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30342287

Brand: Toronto Research Chemicals TRCB6841975G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromoethane-D5
CAS 3675-63-6
Purity Grade Notes GC
Molecular Formula C2 2H5 Br
Quantity 5 g
Synonym Ethane-1,1,1,2,2-d5, 2-bromo- (ACI), 2-Bromoethane-1,1,1,2,2-d5 (ACI), Ethane-d5, bromo- (6CI, 7CI, 8CI, 9CI), 1-Bromo-1,1,2,2,2-pentadeuterioethane, Bromoethane-d5, Ethyl bromide-d5, Ethyl-d5 bromide, Pentadeuterioethyl bromide, Pentadeuteroethyl bromide, Perdeuterioethyl bromide, Bromoethane-D5
InChI Formula InChI=1S/C2H5Br/c1-2-3/h2H2,1H3/i1D3,2D2
SMILES [2H]C([2H])([2H])C([2H])([2H])Br
IUPAC Name 1-bromo-1,1,2,2,2-pentadeuterioethane
Molecular Weight (g/mol) 114
Formula Weight 112.9888
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.