missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldicarb Sulfone, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Aldicarb-sulfone |
| CAS | 1646-88-4 |
| Molecular Formula | C7 H14 N2 O4 S |
| Quantity | 5 mg |
| Synonym | Propionaldehyde, 2-methyl-2-(methylsulfonyl)-, O-(methylcarbamoyl)oxime (7CI,8CI), Carbamic acid, methyl-, O-[[2-methyl-2-(methylsulfonyl)propylidene]amino] deriv. (8CI), 2-Methyl-2-(methylsulfonyl)propionaldehyde O-(methylcarbamoyl)oxime, Aldicarb sulfone, Aldoxycarb, Standak, Sulfocarb, Temik sulfone, UC 21865, UCES, Yangdimiewei |
| InChI Formula | InChI=1S/C7H14N2O4S/c1-7(2,14(4,11)12)5-9-13-6(10)8-3/h5H,1-4H3,(H,8,10) |
| SMILES | CNC(=O)ON=CC(C)(C)S(=O)(=O)C |
| IUPAC Name | [(2-methyl-2-methylsulfonylpropylidene)amino] N-methylcarbamate |
| Molecular Weight (g/mol) | 222.26 |
| Formula Weight | 222.0674 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction