missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzidine-d8, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Benzidine ring-d8 |
| CAS | 92890-63-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 2H8 H4 N2 |
| Quantity | 5 mg |
| Synonym | [1,1'-Biphenyl-2,2',3,3',5,5',6,6'-d8]-4,4'-diamine, Benzidine D8, 4,4'-Diaminobiphenyl-d8, 4,4'-Diamino(biphenyl-d8) |
| InChI Formula | InChI=1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2/i1D,2D,3D,4D,5D,6D,7D,8D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1N)c2c([2H])c([2H])c(N)c([2H])c2[2H] |
| IUPAC Name | 4-(4-amino-2,3,5,6-tetradeuteriophenyl)-2,3,5,6-tetradeuterioaniline |
| Molecular Weight (g/mol) | 192.29 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction