missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-d7 Paraben, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Benzyl-d7 Paraben |
| CAS | 1216916-40-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 D7 H5 O3 |
| Quantity | 5 mg |
| Synonym | 4-Hydroxybenzoic Acid Phenylmethyl-d7 Ester, 4-(Benzyloxy-d7-carbonyl)phenol, 4-Hydroxybenzoic Acid Benzyl-d7 Ester, Benzyl-d7 4-Hydroxybenzoate, Benzyl-d7 4-Hydroxylbenzoate, Benzyl-d7 Parasept, Benzyl-d7 Tegosept-d7, NSC 8080-d7, Nipabenzyl-d7, POB-BZ-d7, Parosept-d7, Solbrol Z-d7 |
| InChI Formula | InChI=1S/C14H12O3/c15-13-8-6-12(7-9-13)14(16)17-10-11-4-2-1-3-5-11/h1-9,15H,10H2/i1D,2D,3D,4D,5D,10D2 |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])OC(=O)c2ccc(O)cc2 |
| IUPAC Name | [dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl] 4-hydroxybenzoate |
| Molecular Weight (g/mol) | 235.29 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction