missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brexpiprazole-d8, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Brexpiprazole-d8 |
| CAS | 1427049-19-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C25H19D8N3O2S |
| Quantity | 5 mg |
| Synonym | 7-[4-(4-(Benzo[b]thien-4-yl)-piperazin-1-yl)butoxy]-1H-quinolin-2-one-d8, OPC 34712-d8, 2(1H)-Quinolinone, 7-[4-(4-benzo[b]thien-4-yl-1-piperazinyl)butoxy-1,1,2,2,3,3,4,4-d8]-, 7-[4-(4-Benzo[b]thien-4-yl-1-piperazinyl)butoxy-1,1,2,2,3,3,4,4-d8]-2(1H)-quinolinone, 7-[4-(4-(Benzo[b]thien-4-yl)-piperazin-1-yl)-1,1,2,2,3,3,4,4-octadeuteriobutoxy]-1H-quinolin-2-one, Brexpiprazole-D8 (butoxy-D8) |
| InChI Formula | InChI=1S/C25H27N3O2S/c29-25-9-7-19-6-8-20(18-22(19)26-25)30-16-2-1-11-27-12-14-28(15-13-27)23-4-3-5-24-21(23)10-17-31-24/h3-10,17-18H,1-2,11-16H2,(H,26,29)/i1D2,2D2,11D2,16D2 |
| SMILES | [2H]C([2H])(Oc1ccc2C=CC(=O)Nc2c1)C([2H])([2H])C([2H])([2H])C([2H])([2H])N3CCN(CC3)c4cccc5sccc45 |
| IUPAC Name | 7-[4-[4-(1-benzothiophen-4-yl)piperazin-1-yl]-1,1,2,2,3,3,4,4-octadeuteriobutoxy]-1H-quinolin-2-one |
| Molecular Weight (g/mol) | 441.62 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction