missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetylpiperazine-N’-(4-phenol) Ketoconazole, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-Acetylpiperazine-N'-(4-phenol) Ketoconazole |
| CAS | 1346598-39-7 |
| Molecular Formula | C38 H42 Cl2 N6 O6 |
| Quantity | 5 mg |
| Synonym | N-Acetylpiperazine-N'-(4-phenol) Ketoconazole |
| InChI Formula | InChI=1S/C38H42Cl2N6O6/c1-27(47)43-13-17-45(18-14-43)30-4-7-32(8-5-30)51-37-22-31(46-19-15-44(16-20-46)28(2)48)6-10-36(37)49-23-33-24-50-38(52-33,25-42-12-11-41-26-42)34-9-3-29(39)21-35(34)40/h3-12,21-22,26,33H,13-20,23-25H2,1-2H3/t33-,38-/m1/s1 |
| SMILES | CC(=O)N1CCN(CC1)c2ccc(Oc3cc(ccc3OC[C@@H]4CO[C@](Cn5ccnc5)(O4)c6ccc(Cl)cc6Cl)N7CCN(CC7)C(=O)C)cc2 |
| IUPAC Name | 1-[4-[4-[5-(4-acetylpiperazin-1-yl)-2-[[(2S,4R)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenoxy]phenyl]piperazin-1-yl]ethanone |
| Molecular Weight (g/mol) | 749.683 |
| Formula Weight | 748.254 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction