missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-2,4-dibromopyrrole, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-BOC-2,4-dibromopyrrole |
| CAS | 117657-38-2 |
| Molecular Formula | C9H11Br2NO2 |
| Quantity | 5 mg |
| Synonym | 2,5-Dibromo-1H-pyrrole-1-carboxylic Acid 1,1-Dimethylethyl Ester, 2,5-Dibromo-1H-pyrrole-1-carboxylic Acid tert-Butyl Ester |
| InChI Formula | InChI=1S/C9H11Br2NO2/c1-9(2,3)14-8(13)12-6(10)4-5-7(12)11/h4-5H,1-3H3 |
| SMILES | CC(C)(C)OC(=O)n1c(Br)ccc1Br |
| IUPAC Name | tert-butyl 2,5-dibromopyrrole-1-carboxylate |
| Molecular Weight (g/mol) | 325 |
| Formula Weight | 322.9157 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction