missing translation for 'onlineSavingsMsg'
Learn More

6-Acetylpyridine-2-carboxylic acid, 95%, Thermo Scientific™

Catalog Number 15406977
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
122637-39-2
C8H7NO3
165.148
MFCD09800364
SMFINMWLJMPOES-UHFFFAOYSA-N
6-acetylpicolinic acid, 6-acetyl-2-pyridinecarboxylic acid, 6-acetyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-acetyl, 6-acetylpicolinicacid, pubchem23361, acmc-1bxea, 6-ethanoylpyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-acetyl
15206504
6-acetylpyridine-2-carboxylic acid
CC(=O)C1=NC(=CC=C1)C(=O)O
This item is not returnable. View return policy
122637-39-2
C8H7NO3
165.148
MFCD09800364
SMFINMWLJMPOES-UHFFFAOYSA-N
6-acetylpicolinic acid, 6-acetyl-2-pyridinecarboxylic acid, 6-acetyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-acetyl, 6-acetylpicolinicacid, pubchem23361, acmc-1bxea, 6-ethanoylpyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 6-acetyl
15206504
6-acetylpyridine-2-carboxylic acid
CC(=O)C1=NC(=CC=C1)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 137°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 165.1
Percent Purity 95%
Chemical Name or Material 6-Acetylpyridine-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'