missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-methyl-3-nitropyridine, 95%, Thermo Scientific™

Catalog Number 15407728
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
22280-62-2
C6H7N3O2
153.141
MFCD00053582
BGMZTBKXOFFTBJ-UHFFFAOYSA-N
2-amino-6-methyl-5-nitropyridine, 6-amino-3-nitro-2-picoline, 2-amino-5-nitro-6-methylpyridine, 6-amino-2-methyl-3-nitropyridine, 2-amino-5-nitro-6-methyl pyridine, 2-amino-5-nitro-6-picoline, 6-amino-3-nitro-2-methylpyridine, 6-methyl-5-nitro-2-pyridinamine, 6-methyl-5-nitro-pyridin-2-ylamine, 6-methyl-5-nitro-2-pyridylamine
247976
6-methyl-5-nitropyridin-2-amine
CC1=C(C=CC(=N1)N)[N+](=O)[O-]
This item is not returnable. View return policy
22280-62-2
C6H7N3O2
153.141
MFCD00053582
BGMZTBKXOFFTBJ-UHFFFAOYSA-N
2-amino-6-methyl-5-nitropyridine, 6-amino-3-nitro-2-picoline, 2-amino-5-nitro-6-methylpyridine, 6-amino-2-methyl-3-nitropyridine, 2-amino-5-nitro-6-methyl pyridine, 2-amino-5-nitro-6-picoline, 6-amino-3-nitro-2-methylpyridine, 6-methyl-5-nitro-2-pyridinamine, 6-methyl-5-nitro-pyridin-2-ylamine, 6-methyl-5-nitro-2-pyridylamine
247976
6-methyl-5-nitropyridin-2-amine
CC1=C(C=CC(=N1)N)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 185°C to 189°C
Color Yellow
Quantity 5 g
Formula Weight 153.14
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6-Amino-2-methyl-3-nitropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'