missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Bromo-4-methylquinoline, 96%

Brand:  Alfa Aesar™ H33298.03

Additional Details : CAS : 41037-28-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15406117

  • 117.00€ / 1g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

41037-28-9
222.085
XBZQSJLYRMZRSZ-UHFFFAOYSA-N
12475362
CC1=C2C=C(C=CC2=NC=C1)Br
C10H8BrN
MFCD09258893
quinoline, 6-bromo-4-methyl, 6-bromo-4-methyl-quinoline, 6-bromo lepidine, 4-methyl-6-bromoquinoline, quinoline,6-bromo-4-methyl, 6-bromanyl-4-methyl-quinoline
6-bromo-4-methylquinoline

Specifications

6-Bromo-4-methylquinoline
94°C to 95°C
41037-28-9
MFCD09258893
XBZQSJLYRMZRSZ-UHFFFAOYSA-N
6-bromo-4-methylquinoline
12475362
96%
Air and light sensitive
1g
C10H8BrN
quinoline, 6-bromo-4-methyl, 6-bromo-4-methyl-quinoline, 6-bromo lepidine, 4-methyl-6-bromoquinoline, quinoline,6-bromo-4-methyl, 6-bromanyl-4-methyl-quinoline
CC1=C2C=C(C=CC2=NC=C1)Br
222.085
222.08
Videos
SDS Documents