missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromopiperonal, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.
Solubility
Soluble in DMSO, methanol
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.
Solubility
Soluble in DMSO, methanol
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 127°C to 130°C |
Quantity | 5 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in DMSO,methanol |
IUPAC Name | 6-bromo-1,3-benzodioxole-5-carbaldehyde |
Formula Weight | 229.03 |
Percent Purity | 98% |
Chemical Name or Material | 6-Bromopiperonal |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'