missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluoro-DL-Tryptophan, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 6-Fluoro-DL-Tryptophan |
| CAS | 7730-20-3 |
| Molecular Formula | C11H11FN2O2 |
| Quantity | 100 mg |
| Synonym | 6-Fluorotryptophan, (±)-6-Fluorotryptophan, DL-6-Fluorotryptophan, DL-6-Fluorotryptophane, NSC 9364 |
| InChI Formula | InChI=1S/C11H11FN2O2/c12-7-1-2-8-6(3-9(13)11(15)16)5-14-10(8)4-7/h1-2,4-5,9,14H,3,13H2,(H,15,16) |
| SMILES | FC1=CC2=C(C(CC(N)C(O)=O)=CN2)C=C1 |
| Molecular Weight (g/mol) | 222.2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction