missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorosalicylic acid, 96%, Thermo Scientific™

Product Code. 11363066
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11363066

Brand: Thermo Scientific Alfa Aesar L10907.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67531-86-6
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.112
MDL Number MFCD00052179
InChI Key BCEKGWWLVKXZKK-UHFFFAOYSA-N
Synonym 6-fluorosalicylic acid, 6-fluoro-2-hydroxybenzoic acid, 2-fluoro-6-hydroxybenzoicacid, 6-fluorosalicylicacid, benzoic acid, 2-fluoro-6-hydroxy, 2-fluoro-6-hydoxybenzoic acid, pubchem4958, acmc-1baav, 2-carboxy-3-fluorophenol
PubChem CID 586777
IUPAC Name 2-fluoro-6-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)F)C(=O)O)O

Specifications

Melting Point 159°C to 161°C
Quantity 250 mg
Beilstein 7111497
Formula Weight 156.11
Percent Purity 96%
Chemical Name or Material 6-Fluorosalicylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.