missing translation for 'onlineSavingsMsg'
Learn More

6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine, 97%, Thermo Scientific™

Catalog Number p-2703369
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
175202-00-3
C9H13N5
191.24
MFCD00067969
NDFXPLOECYNSHX-UHFFFAOYSA-N
6-hydrazino-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethylpyrazolo 3,4-b pyridine, 1,3,4-trimethylpyrazolo 5,4-b pyridine-6-ylhydrazine, 1,3,4-trimethylpyrazolo 3,4-b pyridin-6-yl hydrazine, 1h-pyrazolo 3,4-b pyridine,6-hydrazinyl-1,3,4-trimethyl, 6h-pyrazolo 3,4-b pyridin-6-one, 1,7-dihydro-1,3,4-trimethyl-, hydrazone 9ci
2778646
6-hydrazinyl-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
CN1N=C(C)C2=C(C)C=C(NN)N=C12
This item is not returnable. View return policy
175202-00-3
C9H13N5
191.24
MFCD00067969
NDFXPLOECYNSHX-UHFFFAOYSA-N
6-hydrazino-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethylpyrazolo 3,4-b pyridine, 1,3,4-trimethylpyrazolo 5,4-b pyridine-6-ylhydrazine, 1,3,4-trimethylpyrazolo 3,4-b pyridin-6-yl hydrazine, 1h-pyrazolo 3,4-b pyridine,6-hydrazinyl-1,3,4-trimethyl, 6h-pyrazolo 3,4-b pyridin-6-one, 1,7-dihydro-1,3,4-trimethyl-, hydrazone 9ci
2778646
6-hydrazinyl-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
CN1N=C(C)C2=C(C)C=C(NN)N=C12
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 191.24
Percent Purity 97%
Chemical Name or Material 6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'