missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyindole, 98%, Thermo Scientific Chemicals

Catalog Number 15412358
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2380-86-1
C8H7NO
133.15
MFCD00152101
XAWPKHNOFIWWNZ-UHFFFAOYSA-N
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol
524508
1H-indol-6-ol
C1=CC(=CC2=C1C=CN2)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2380-86-1
C8H7NO
133.15
MFCD00152101
XAWPKHNOFIWWNZ-UHFFFAOYSA-N
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol
524508
1H-indol-6-ol
C1=CC(=CC2=C1C=CN2)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 125°C to 128°C
Quantity 5 g
UN Number UN3077
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 6-Hydroxyindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'