missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroindole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7054654
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
10242-00-9
C9H6N2O4
206.157
MFCD02664466
JIXJNWVLWQQNDB-UHFFFAOYSA-N
6-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-nitro, 6-nitro-2-indolecarboxylic acid, 1h-indole-2-carboxylicacid, 6-nitro, 6-nitroindol-2-carboxylic acid
4714997
6-nitro-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1[N+](=O)[O-])NC(=C2)C(=O)O
This item is not returnable. View return policy
10242-00-9
C9H6N2O4
206.157
MFCD02664466
JIXJNWVLWQQNDB-UHFFFAOYSA-N
6-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-nitro, 6-nitro-2-indolecarboxylic acid, 1h-indole-2-carboxylicacid, 6-nitro, 6-nitroindol-2-carboxylic acid
4714997
6-nitro-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1[N+](=O)[O-])NC(=C2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 206.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Nitroindole-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'