missing translation for 'onlineSavingsMsg'
Learn More

Consort 6010 Multiparameter Portable Analyzer

Product Code. p-7147294
Click to view available options
Product Type:
Analyzer with pH Kit
Portable Analyzer
Unit Size:
Each
This item is not returnable. View return policy
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.

Product Code. 12117810

Brand: Consort C6010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.

Suitable as bench-top and as portable device. Consort 6010 Multiparameter Portable Analyzer is a single-channel meter compatible with an array of electrodes that features an easy-to-use menu system and built-in help system to guide users through configuration and use.

Meter available separately or with pH kit (pH/ATC electrode SP20T, 2 x 50mL buffers (pH 4 and 7), 50mL electrolyte (3M KCl), and carrying case).

  • Portable and bench-top combined in one instrument
  • Parameters: pH, mV, conductivity, resistivity, salinity, TDS, dissolved oxygen, temperature
  • Gold plated BNC connectors resist corrosion to ensure a long life, even in harsh environments
  • Pre-programmed pH buffers and EC standards, plus add up to 5 pH buffers and 3 EC standards
  • Custom calibration tables provide complete buffer/standard-temperature relation info (add via menu or from PC)
  • Stability indicator ensures visualization when the measurement has stabilized
  • Stability algorithm ensures stable readings with ability to detect fast changes
  • Hold function allows to freeze the display
  • Selectable resolution for more stable readings on mV, pH and DO
  • Range lock for conductivity measurements
  • Capacitive compensation eliminates electrode/cable capacitive component with low conductivities
  • Galvanic isolated USB interface eliminates ground loop effects when connected to a PC
  • GPL report can be shown on the display or sent to the digital port
  • Pre-programmed standards
    pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 (at 25°C)
    Conductivity: 1413 μS/cm, 12.88 mS/cm, 111.8 mS/cm (at 25°C)
  • Free software and firmware updates downloadable from www.consort.be

Spécification

Outputs USB
Accuracy (Conductivity) 0.5% f.s. of range
Accuracy (DO) 1% ± 1 digit
Accuracy (mV) 0.1% ± 1 digit
Accuracy (pH) 0.1% ±1 digit
Backlight Option White backlight
Battery Type 4 x 1.2 V, AA, NiMH
Calibration GLP, reminder: 0 - 999 h
Calibration Points 1 (mV, DO, air pressure)
Calibration Points (Conductivity) 1 to 3
Calibration Points (pH) 1 to 5
Cell Constant 0.07 to 13 cm-1
Data Management Details All modes, manual or timed, interval: 1 - 9999 s
Data Points 12,000 + °C/date/time
Channels 1 measurement, 1 temperature
Display Type LCD, 128 x 64 pixel
Range (Conductivity) 0 to 2000 mS/cm
Range (mV) ±2000.0
Range (pH) -2.000 to 16.000
Range (Resistivity) 0 to 200 Mohm/cm
Range (Salinity) 0.0 to 70.0
Range (TDS) 0 to 100.0 g/L
Resolution (pH) 0.001
Temperature Range (Metric) -5°C to +105°C
Includes Meter Only
Inputs 1 BNC for measurement, 1 x 2 banana for temp
Languages English, Dutch, German, French
Model C6010
Power Supply 100/240 VAC, 50/60 Hz
Product Type Portable Analyzer
Range 600 to 1300 hPa (air pressure)
Range (RDO) 0 to 60.00 mg/L
Relative Humidity Range 0 - 95%, non-condensing
Resolution (Conductivity) 0.001 μS/cm
Resolution (mV) 0.1
Resolution (RDO) 0.01 mg/L
Resolution (Resistivity) 1 Ω.cm
Resolution (TDS) 0.01 mg/L
Temperature Accuracy 0.1°C
Temperature Compensation −5° to +105° C (pH, conductivity), 0° to 50° C (DO)
Temperature (Metric) Operating 0°C to 40°C
Temperature Probe Calibration 1 point
Temperature Resolution 0.1° C
Temperature Sensor Pt1000
Warranty 36 Months
Dimensions (L x W x H) 12W x 25D x 5H cm
Weight (Metric) 600 g
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis