Learn More
YSI™ 626250 ProODO Cable and Probe
Product Code. 11958118
Click to view available options
Length (Metric) Cable:
1 m
10 m
4 m
50 m
Length (English) Cable:
3 ft.
13 ft.
32 ft.
164 ft.
Includes:
Sensing cap
Sensing cap and cable management kit
Unit Size:
Each
Product Code. 11958118
Brand: YSI™ 6262501
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Stores calibration data within the sensor, allowing probes to be placed on any ProODO instrument without re-calibration
For use with ProODO Handheld Meters exclusively.
  • Assemblies include a ProODO optical DO sensor and sensing cap
  • Cable assemblies over one meter also include a cable management kit
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±1%mg/L
Length (English) Cable 3 ft.
Length (Metric) Cable 1 m
Temperature (Metric) Range 10°C to 40°C
Certifications/Compliance RoHS, CE, WEEE, IP65
Includes Sensing cap
Temperature Resolution 0.1°C
Warranty 2 Years
For Use With (Equipment) DO/temperature meter
Probe Type ProODO
For Use With (Application) Stores calibration data within the sensor