Learn More
YSI™ 626250 ProODO Cable and Probe
Product Code. 11978118
Click to view available options
Length (Metric) Cable:
1 m
10 m
4 m
50 m
Length (English) Cable:
3 ft.
13 ft.
32 ft.
164 ft.
Includes:
Sensing cap
Sensing cap and cable management kit
Unit Size:
Each
Product Code. 11978118
Brand: YSI™ 62625010
Only null left
Add to basket
Add to basket
Stores calibration data within the sensor, allowing probes to be placed on any ProODO instrument without re-calibration
For use with ProODO Handheld Meters exclusively.
  • Assemblies include a ProODO optical DO sensor and sensing cap
  • Cable assemblies over one meter also include a cable management kit
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±1%mg/L
Length (English) Cable 32 ft.
Length (Metric) Cable 10 m
Temperature (Metric) Range 10°C to 40°C
Certifications/Compliance RoHS, CE, WEEE, IP65
Includes Sensing cap and cable management kit
Temperature Resolution 0.1°C
Warranty 2 Years
For Use With (Equipment) DO/temperature meter
Probe Type ProODO
For Use With (Application) Stores calibration data within the sensor