missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CSMD3 Protein

Product Code. 18763969
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18763969

Brand: R&D Systems™ 9689CS050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Recombinant Human CSMD3 Protein CF is a new member of CSMD (CUB and sushi multiple domains) gene family.

The Recombinant Human CSMD3 Protein is derived from CHO.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7Z407-1
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 114788
Molecular Weight (g/mol) 119 kDa
Name CSMD-3, CUB and sushi domain-containing protein 3, CUB and Sushi multiple domains 3, KIAA1894, KIAA1894CUB and sushi multiple domains protein 3
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human CSMD3 protein Ala2218-Ser3306,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.