missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Bromo-1-benzofuran, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 7-Bromo-1-benzofuran |
| CAS | 133720-60-2 |
| Molecular Formula | C8H5BrO |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C8H5BrO/c9-7-3-1-2-6-4-5-10-8(6)7/h1-5H |
| SMILES | Brc1cccc2ccoc12 |
| IUPAC Name | 7-bromo-1-benzofuran |
| Molecular Weight (g/mol) | 197.03 |
| Formula Weight | 195.9524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction