Learn More
7-Bromoheptanoic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-7077411
Click to view available options
Quantity:
1 g
5 g
25 g
30515-28-7
C7H13BrO2
209.08
JLPQXFFMVVPIRW-UHFFFAOYSA-N
7-bromo-heptanoic acid, heptanoic acid, 7-bromo, 7-bromoenanthic acid, omega-bromoheptanoic acid, 7-bromoheptanoicacid, 5-bromopentylacetate;, 7-bromo heptanoic acid, acmc-1aezo, ksc226m8n
121723
7-bromoheptanoic acid
C(CCCBr)CCC(=O)O
30515-28-7
C7H13BrO2
209.08
JLPQXFFMVVPIRW-UHFFFAOYSA-N
7-bromo-heptanoic acid, heptanoic acid, 7-bromo, 7-bromoenanthic acid, omega-bromoheptanoic acid, 7-bromoheptanoicacid, 5-bromopentylacetate;, 7-bromo heptanoic acid, acmc-1aezo, ksc226m8n
121723
7-bromoheptanoic acid
C(CCCBr)CCC(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 29.0°C to 32.0°C
Boiling Point 121.0°C to 125.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 209.08
Percent Purity 97%
Chemical Name or Material 7-Bromoheptanoic acid
compliance-icons
Product Identifier
  • 7-Bromoheptanoic acid, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.