missing translation for 'onlineSavingsMsg'
Learn More

7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Thermo Scientific™

Catalog Number p-2703532
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
306934-90-7
C9H9IO2
276.073
MFCD02677706
CIZHAPZOOAJDCN-UHFFFAOYSA-N
7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
2776396
7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
C1COC2=C(C=C(C=C2)I)OC1
This item is not returnable. View return policy
306934-90-7
C9H9IO2
276.073
MFCD02677706
CIZHAPZOOAJDCN-UHFFFAOYSA-N
7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
2776396
7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
C1COC2=C(C=C(C=C2)I)OC1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 276.07
Percent Purity 95%
Chemical Name or Material 7-Iodo-3, 4-dihydro-2H-1, 5-benzodioxepine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'