Learn More
Alfa Aesar™ 7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine, 95%
Brand: Alfa Aesar™ H34168.03
Additional Details : CAS : 274676-47-0 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
274676-47-0 | |
148.209 | |
HBWCJMBLUCNJIS-UHFFFAOYSA-N | |
22319466 | |
CC1=NC2=C(CCCN2)C=C1 |
C9H12N2 | |
MFCD06739289 | |
2-methyl-5,6,7,8-tetrahydro-1,8-naphthyridine, 7-methyl-1,2,3,4-tetrahydro-1,8 naphthyridine, 7-methyl-1,2,3,4-tetrahydropyridino 2,3-b pyridine, 1,2,3,4-tetrahydro-7-methyl-1,8-naphthyridine, 1,8-naphthyridine, 1,2,3,4-tetrahydro-7-methyl-9ci | |
7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine |
Specifications
7-Methyl-1,2,3,4-tetrahydro-1,8-naphthyridine | |
1g | |
C9H12N2 | |
2-methyl-5,6,7,8-tetrahydro-1,8-naphthyridine, 7-methyl-1,2,3,4-tetrahydro-1,8 naphthyridine, 7-methyl-1,2,3,4-tetrahydropyridino 2,3-b pyridine, 1,2,3,4-tetrahydro-7-methyl-1,8-naphthyridine, 1,8-naphthyridine, 1,2,3,4-tetrahydro-7-methyl-9ci | |
CC1=NC2=C(CCCN2)C=C1 | |
148.209 | |
148.21 |
88°C to 91°C | |
274676-47-0 | |
MFCD06739289 | |
HBWCJMBLUCNJIS-UHFFFAOYSA-N | |
7-methyl-1,2,3,4-tetrahydro-1,8-naphthyridine | |
22319466 | |
95% |