Learn More
SHOWA 707HVO Orange Nitrile Gloves
Product Code. 17897311
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 17897311
Brand: SHOWA 707HVO06XS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The 707HVO gloves are highly tactile and fit like a second skin, all whilst protecting against grease, chemicals, and abrasion.
  • Fluorescent
  • Lightweight
  • Chlorinated
  • Eco Best Technology (EBT)
  • Unlined
  • Unsupported
Specifications
Certifications/Compliance (Europe) EN 388:2016, EN ISO 374-1:2016/Type A, EN ISO 374-5:2016
Color Orange
Size 6
Glove Cuff Style Rolled
Model 707HVO