missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoquinoline, 98+%, Thermo Scientific Chemicals

Catalog Number p-3735511
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
578-66-5
C9H8N2
144.18
MFCD00006809
WREVVZMUNPAPOV-UHFFFAOYSA-N
8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
11359
quinolin-8-amine
C1=CC2=C(C(=C1)N)N=CC=C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
578-66-5
C9H8N2
144.18
MFCD00006809
WREVVZMUNPAPOV-UHFFFAOYSA-N
8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
11359
quinolin-8-amine
C1=CC2=C(C(=C1)N)N=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 62.0°C to 65.0°C
Color Beige-Brown to Green
Boiling Point 174.0°C (26.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22, 450
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, insoluble in ether
Formula Weight 144.18
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 8-Aminoquinoline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'