missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Aminophenanthrene, 96%

Brand:  Alfa Aesar™ H55773.03

Additional Details : CAS : 947-73-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412128

  • 586.00€ / 1g


Estimated Shipment
17-05-2021
Add to basket

Description

Description

Applications
9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

Solubility
It is slightly soluble in water 0.00581 mg/mL.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

947-73-9
193.249
KIHQWOBUUIPWAN-UHFFFAOYSA-N
13695
phenanthren-9-amine
C14H11N
MFCD00001177
9-aminophenanthrene, 9-phenanthrenamine, 9-phenanthrylamine, phenanthren-9-ylamine, unii-7m9m5km2xy, ccris 7006, 7m9m5km2xy, 9ap, acmc-20alvr, phenanthren-9-yl-amine
CHEBI:50475
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)N

Specifications

9-Aminophenanthrene
137°C to 139°C
947-73-9
MFCD00001177
It is slightly soluble in water 0.00581mg/mL.
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)N
193.249
CHEBI:50475
Solid
White to Brown
1g
C14H11N
9-aminophenanthrene, 9-phenanthrenamine, 9-phenanthrylamine, phenanthren-9-ylamine, unii-7m9m5km2xy, ccris 7006, 7m9m5km2xy, 9ap, acmc-20alvr, phenanthren-9-yl-amine
KIHQWOBUUIPWAN-UHFFFAOYSA-N
phenanthren-9-amine
13695
193.24
96%
Videos
SDS Documents