Learn More
9-beta-D-Arabinofuranosyladenine, 99%, Thermo Scientific Chemicals
Description
Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 ?M. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Vidarabine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes. Vidarabine is an inhibitor of A cyclase and DNA Polymerase. Used to study the roles of AMP-activated protein kinase (AMPK) in cell signaling. Cell-permeable adenylate cyclase inhibitor; in detergent-dispersed rat brain preparation, IC50 = 30 μM. Clinically significant antiviral agent, especially against herpes simplex (HSV),1 by inhibition of DNA polymerase.
Solubility
Soluble in DMF (10 mg/ml), 0.5 M HCl (50 mg/ml), DMSO (53 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), and water (3 mg/ml at 25°C).
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Color | White |
Quantity | 5 g |
Beilstein | 624881 |
Merck Index | 14,9976 |
Solubility Information | Soluble in DMF (10mg/ml),0.5 M HCl (50mg/ml),DMSO (53mg/ml at 25°C),ethanol (<1mg/ml at 25°C),and water (3mg/ml at 25°C). |
Formula Weight | 267.24 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | 9-beta-D-Arabinofuranosyladenine |
RUO – Research Use Only