missing translation for 'onlineSavingsMsg'
Learn More

9-Decen-1-ol, 90+%

Product Code. 11434067
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11434067

Brand: Thermo Scientific Alfa Aesar A19490.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13019-22-2 | C10H20O | 156.269 g/mol

It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

Solubility
Soluble in water at 20°C 0.16g/L. Soluble in alcohol, dipropylene glycol, paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13019-22-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002992
InChI Key QGFSQVPRCWJZQK-UHFFFAOYSA-N
Synonym 9-decen-1-ol, decylenic alcohol, 1-decen-10-ol, omega-decenol, 9-decenol, omega-decen-1-ol, .omega.-decen-1-ol, .omega.-decenol, 9-decene-1-ol, 9-decene1-ol
PubChem CID 25612
IUPAC Name dec-9-en-1-ol
SMILES C=CCCCCCCCCO

Specifications

Melting Point -13°C
Density 0.847
Boiling Point 235°C
Flash Point 104°C (219°F)
Odor Pleasant
Refractive Index 1.448
Quantity 100 g
Beilstein 1750928
Solubility Information Soluble in water at 20°C 0.16g/L. Soluble in alcohol,dipropylene glycol,paraffin oil.
Formula Weight 156.27
Percent Purity ≥90%
Chemical Name or Material 9-Decen-1-ol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.