missing translation for 'onlineSavingsMsg'
Learn More

Uvex 94907 Comfortable Climate Insole For Uvex G2 - Width 10

Comfortable climatic insole with antistatic properties for Uvex 1 G2

POA

Specifications

Color Black
For Use With (Equipment) Uvex G2
Material Microvelour
Disposable No
Gender Unisex
View More Specs

Description

Description

  • Full-surface foot shock absorption
  • Highly breathable, moisture-absorbent and quickly re-drying material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No: 9490.7
  • Sizes: 35 to 52
Specifications

Specifications

Black
Microvelour
Unisex
Yes
Uvex G2
No
Narrow Width
No
Videos
SDS
Documents

Documents

Certificates