missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ 96-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Brand: Greiner Bio-One 655946
Additional Details : Weight : 1.35000kg
Description
- Poly-D-Lysine (PDL) are synthetic molecules that are used to improve the adhesion of different cell types to polystyrene surfaces
- Shelf life: 18 months at room temperature
- Molecular weight PDL: 75 - 150 kD
- Promotion of cell adhesion, proliferation and growth of transfected cell lines (e.g. HEK-293, PC12, L929, certain 3T3 cell lines), neuronal cell lines, as well as primary neurons and glia cells
- Chimey well design allows each individual well to stand on its own minimizing the risk of sample carryover and cross contamination
Advantages:
- Increase in isolation and cultivation efficiency
- Ready-to-use products: immediate use, time-saving
- Consistent quality
- Storable at room temperature
Specifications
Alphanumeric | |
Standard | |
34 mm2 | |
5/Pk., 20/Cs. | |
Polystyrene | |
Non-sterile | |
392 μL | |
Yes |
ANSI (ANSI/SBS 1-2004) | |
With Lid | |
8 x 12 | |
Black, Clear Bottom | |
96 | |
Poly-D-Lysine | |
Flat Bottom (F-well) | |
20/Cs. |