missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9954 Spectacle Storage
Click to view available options
Product Type:
Goggle Case
Microfiber Spectacle Bag
Description
- For use with Uvex™ Safety Spectacles and Goggles
Specifications
Color | Black |
Product Type | Goggle Case |
For Use With (Equipment) | Uvex™ Safety Goggles |
For Use With (Application) | Storing/Transporting Eyewear |
Disposable | Reusable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'