missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethylbenzene, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 012411.B4

Product Code. 11311789

  • 543.00€ / 16kg
 Restricted Item
Estimated Shipment: 14-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

95-63-6
169°C to 171°C
Aromatic
1.504
16kg
1903005
pseudocumene, pseudocumol, psi-cumene, as-trimethylbenzene, 1,3,4-trimethylbenzene, benzene, 1,2,4-trimethyl, uns-trimethylbenzene, 1,2,5-trimethylbenzene, asymmetrical trimethylbenzene, .psi.-cumene
GWHJZXXIDMPWGX-UHFFFAOYSA-N
1,2,4-trimethylbenzene
7247
120.2
1,2,4-Trimethylbenzene
0.882 g/mL
48°C (118°F)
C9H12
MFCD00008527
UN3295
14,7915
Soluble in alcohol,benzene and ether; Slightly soluble in water
CC1=CC(=C(C=C1)C)C
120.195
CHEBI:34039
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.