missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-hydroxyanthraquinone, 96%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar B24388.22

Quantity 100g

Product Code. 11455827

  • 298.86€ / 100g
 Restricted Item
Estimated Shipment: 02-12-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

Solubility
Soluble in water, ethanol, acetone, benzene.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

116-85-8
MFCD00001223
1-amino-4-hydroxyanthraquinone, disperse red 15, calcosyn pink b, celutate pink b, nacelan pink b, amacel pink b, celutate pink bn, celutate pink by, cibacete red 3b, disperse red 2s
AQXYVFBSOOBBQV-UHFFFAOYSA-N
1-amino-4-hydroxyanthracene-9,10-dione
8323
96%
C14H9NO3
100g
Soluble in water,ethanol,acetone,benzene.
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N
239.23
239.23
1-Amino-4-hydroxyanthraquinone
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.