missing translation for 'onlineSavingsMsg'
Learn More

Chloroform-d, 99.8% (Isotopic), contains 0.03% v/v TMS, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 041389.AC

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15406375

  • 52.50€ / 25mL
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.50 g/mL
Light and moisture sensitive
865-49-6
MFCD00000827
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
HEDRZPFGACZZDS-MICDWDOJSA-N
trichloro(deuterio)methane
71583
120.38
Chloroform-d, Isotopic
1.4445
25mL
CHCl3
UN1888
Miscible with organic solvents,ethyl acetate and acetone.
C(Cl)(Cl)Cl
120.375
CHEBI:85365
99.8%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.