missing translation for 'onlineSavingsMsg'
Learn More

Praziquantel, 98%, Thermo Scientific™

An anti-helminthic compound

Brand:  Thermo Scientific Alfa Aesar J66188.06

Quantity 5g

Product Code. 15447449

  • 80.80€ / 5g
 Restricted Item
Estimated Shipment: 25-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

Solubility
Freely soluble in ethanol or dichloromethane. Slightly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

55268-74-1
>110°C (230°F)
MFCD00058531
761557
praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum
FSVJFNAIGNNGKK-UHFFFAOYSA-N
2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one
4891
98%
1377°C
C19H24N2O2
5g
14,7716
Freely soluble in ethanol or dichloromethane; Slightly soluble in water
C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2
312.413
312.41
Praziquantel
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.